Bis(4-methoxyphenyl)phenylphosphin

Lieferant: Apollo Scientific
Achtung

Synonyme: Bis(4-methoxyphenyl)(phenyl)phosphine

OR23446-5G
APOSOR23446-5GEA 115 EUR
APOSOR23446-5G
Bis(4-methoxyphenyl)phenylphosphin
Bis(4-methoxyphenyl)phenylphosphin
Formel: C₂₀H₁₉O₂P
Molecular Weight: 322,34 g/mol
Schmelzpunkt: 88…90 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00048993
CAS-Nummer: 14180-51-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR