2-[Di(methylthio)methylidene]malononitrile 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(Bis(methylthio)methylene)malononitrile

OR23452-25G
APOSOR23452-25GEA 132 EUR
APOSOR23452-25G
2-[Di(methylthio)methylidene]malononitrile 98%
2-[Di(methylthio)methylidene]malononitrile
Formel: C₆H₆N₂S₂
Molecular Weight: 170.26 g/mol
Schmelzpunkt: 76…82 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00052730
CAS-Nummer: 5147-80-8
UN: 3439
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR