2-Cyano-N-ethylacetamide
Lieferant: Apollo Scientific
|
Achtung
|
OR23490-1G
APOSOR23490-1GEA
21.9
EUR
APOSOR23490-1G
2-Cyano-N-ethylacetamide
2-Cyano-N-ethylacetamide
Formel:
C₅H₈N₂O Schmelzpunkt: 73…74 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00118415 CAS-Nummer: 15029-36-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...