N-(4-Methyl-1,3-thiazol-2-yl)guanidine hydrochloride
Lieferant: Apollo Scientific
|
Achtung
|
OR23493-1G
APOSOR23493-1GEA
116
EUR
APOSOR23493-1G
N-(4-Methyl-1,3-thiazol-2-yl)guanidine hydrochloride
N-(4-Methyl-1,3-thiazol-2-yl)guanidine hydrochloride
Formel:
C₅H₉ClN₄S Schmelzpunkt: 188…190 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00053024 CAS-Nummer: 100599-91-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...