4-(3-Aminopropyl)-2,4-dihydro-3H-pyrazol-3-one

Lieferant: Apollo Scientific
Gefahr

OR23568-250MG
APOSOR23568-250MGEA 48 EUR
APOSOR23568-250MG
4-(3-Aminopropyl)-2,4-dihydro-3H-pyrazol-3-one
4-(3-Aminopropyl)-2,4-dihydro-3H-pyrazol-3-one
Formel: C₆H₁₁N₃O
Molecular Weight: 141,17 g/mol
Schmelzpunkt: 216 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00053049
CAS-Nummer: 7032-17-9
UN: 3259
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR