2,6-Dichloro-4-methylsulphonylphenol 95+%

Lieferant: Apollo Scientific
Achtung

OR2358-1G
APOSOR2358-1GEA 69.6 EUR
APOSOR2358-1G
2,6-Dichloro-4-methylsulphonylphenol 95+%
2,6-Dichloro-4-methylsulphonylphenol
Formel: C₇H₆Cl₂O₃S
Molecular Weight: 241.09 g/mol
Schmelzpunkt: 223…224 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00216500
CAS-Nummer: 20951-05-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR