6-Chloro-8-(chloromethyl)-4H-1,3-benzodioxine
Lieferant: Apollo Scientific
|
Achtung
|
OR23581-1G
APOSOR23581-1GEA
0
EUR
APOSOR23581-1G
6-Chloro-8-(chloromethyl)-4H-1,3-benzodioxine
6-Chloro-8-(chloromethyl)-4H-1,3-benzodioxine
Formel:
C₉H₈Cl₂O₂ Schmelzpunkt: 99…101 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00053071 CAS-Nummer: 175136-61-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...