1,10-Phenanthrolin Hydrochlorid Monohydrat 97%

Lieferant: Apollo Scientific
Gefahr

OR2360-100G OR2360-250G
APOSOR2360-100GEA 0 EUR
APOSOR2360-100G APOSOR2360-250G
1,10-Phenanthrolin Hydrochlorid Monohydrat 97%
1,10-Phenanthrolin Hydrochlorid Monohydrat
Formel: C₁₂H₉ClN₂·1H₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD00150061
CAS-Nummer: 18851-33-7
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR