4-Methyl-1,2,3-thiadiazole-5-carboxamide

Lieferant: Apollo Scientific
Achtung

OR23651-1G
APOSOR23651-1GEA 66.7 EUR
APOSOR23651-1G
4-Methyl-1,2,3-thiadiazole-5-carboxamide
4-Methyl-1,2,3-thiadiazole-5-carboxamide
Formel: C₄H₅N₃OS
Lagertemperatur: Raumtemperatur
MDL: MFCD00052214
CAS-Nummer: 175136-67-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR