2-Bromo-N-(4-phenyl-1,3-thiazol-2-yl)acetamide

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-[(Bromoacetyl)amino]-4-phenyl-1,3-thiazole, N-(Bromoacetyl)-4-phenyl-1,3-thiazol-2-amine

OR23657-250MG
APOSOR23657-250MGEA 296 EUR
APOSOR23657-250MG
2-Bromo-N-(4-phenyl-1,3-thiazol-2-yl)acetamide
2-Bromo-N-(4-phenyl-1,3-thiazol-2-yl)acetamide
Formel: C₁₁H₉BrN₂OS
Molecular Weight: 297,171 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00178001
CAS-Nummer: 19749-86-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR