2,5-Dibrom-3,4-dinitrothiophen 97%

Lieferant: Apollo Scientific
Achtung

OR23724-5G
APOSOR23724-5GEA 65.4 EUR
APOSOR23724-5G
2,5-Dibrom-3,4-dinitrothiophen 97%
2,5-Dibromo-3,4-dinitrothiophene
Formel: C₄Br₂N₂O₄S
Molecular Weight: 331,94 g/mol
Schmelzpunkt: 134…140 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00015537
CAS-Nummer: 52431-30-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR