2-Iodmesitylen 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Iod-2,4,6-trimethylbenzol, 2-Iod-1,3,5-trimethylbenzol, 2,4,6-Trimethyliodbenzol

OR23747-100G
APOSOR23747-100GEA 132 EUR
APOSOR23747-100G
2-Iodmesitylen 98%
2-Iodmesitylen
Formel: C₉H₁₁I
Molecular Weight: 246,09 g/mol
Schmelzpunkt: 28…30 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00013707
CAS-Nummer: 4028-63-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR