2,6-Dimethyldecane

Lieferant: Apollo Scientific
Gefahr

OR2375-1G
APOSOR2375-1GEA 377 EUR
APOSOR2375-1G
2,6-Dimethyldecane
2,6-Dimethyldecane
Formel: C₁₂H₂₆
Molecular Weight: 170.335 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00060964
CAS-Nummer: 13150-81-7
UN: 1993
ADR: 3 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR