2-[(5-Nitropyridin-2-yl)oxy]ethanol

Lieferant: Apollo Scientific

Synonyme: 2-(2-Hydroxyethoxy)-5-nitropyridine

OR23766-250MG
APOSOR23766-250MGEA 21.9 EUR
APOSOR23766-250MG
2-[(5-Nitropyridin-2-yl)oxy]ethanol
2-[(5-Nitropyridin-2-yl)oxy]ethanol
Formel: C₇H₈N₂O₄
Molecular Weight: 184,15 g/mol
Schmelzpunkt: 111…113 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00052642
CAS-Nummer: 143071-39-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR