1-(4,5-Dihydro-1H-imidazol-2-yl)-3,5-dimethyl-1H-pyrazole hydrobromide

Lieferant: Apollo Scientific
Achtung

Synonyme: 4,5-Dihydro-2-(3,5-dimethyl-1H-pyrazol-1-yl)-1H-imidazole hydrobromide

OR23795-1G
APOSOR23795-1GEA 161 EUR
APOSOR23795-1G
1-(4,5-Dihydro-1H-imidazol-2-yl)-3,5-dimethyl-1H-pyrazole hydrobromide
1-(4,5-Dihydro-1H-imidazol-2-yl)-3,5-dimethyl-1H-pyrazole hydrobromide
Formel: C₈H₁₂N₄·BrH
Schmelzpunkt: 270…275 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00053026
CAS-Nummer: 132369-02-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR