5,6-Dichlor-2-methylbenzimidazol

Lieferant: Apollo Scientific
Achtung

OR2383-5G
APOSOR2383-5GEA 125 EUR
APOSOR2383-5G
5,6-Dichlor-2-methylbenzimidazol
5,6-Dichloro-2-methylbenzimidazole
Formel: C₈H₆Cl₂N₂
Molecular Weight: 201,0500 g/mol
Schmelzpunkt: 248 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00276973
CAS-Nummer: 6478-79-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR