4-Amino-3,5-dimethyl-1,2,4-triazol

Lieferant: Apollo Scientific
Achtung

OR23833-1G OR23833-5G
APOSOR23833-1GEA 72.5 EUR
APOSOR23833-1G APOSOR23833-5G
4-Amino-3,5-dimethyl-1,2,4-triazol
4-Amino-3,5-dimethyl-1,2,4-triazole
Formel: C₄H₈N₄
Molecular Weight: 112.14 g/mol
Schmelzpunkt: 196…198 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00051647
CAS-Nummer: 3530-15-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR