(2-Chlorphenyl)aceton
Lieferant: Apollo Scientific
|
Achtung
|
OR23885-5G
APOSOR23885-5GEA
0
EUR
APOSOR23885-5G
(2-Chlorphenyl)aceton
(2-Chlorphenyl)aceton
Formel:
C₉H₉ClO Siedepunkt: 102…103 °C/3mmHg Lagertemperatur: Raumtemperatur |
MDL:
MFCD00045175 CAS-Nummer: 6305-95-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...