(4-Chlorophenyl)(piperidin-4-yl)methanone hydrochloride

Lieferant: Apollo Scientific
Achtung

OR23895-250MG
APOSOR23895-250MGEA 40.6 EUR
APOSOR23895-250MG
(4-Chlorophenyl)(piperidin-4-yl)methanone hydrochloride
(4-Chlorophenyl)(piperidin-4-yl)methanone hydrochloride
Formel: C₁₂H₁₅Cl₂NO
Molecular Weight: 260,1600 g/mol
Schmelzpunkt: 238…239 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00053028
CAS-Nummer: 55695-51-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR