Ethyl-2-((dimethylamino)methylene)-3-oxobutanoate

Lieferant: Apollo Scientific
Achtung

OR23897-1G OR23897-5G
APOSOR23897-1GEA 0 EUR
APOSOR23897-1G APOSOR23897-5G
Ethyl-2-((dimethylamino)methylene)-3-oxobutanoate
Ethyl-2-((dimethylamino)methylene)-3-oxobutanoate
Formel: C₉H₁₅NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00087563
CAS-Nummer: 51145-57-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR