Ethyl-2-amino-4-methylpyrimidine-5-carboxylate 97%

Lieferant: Apollo Scientific

Synonyme: Ethyl 2-amino-4-methylpyrimidine-5-carboxylate

OR23898-1G
APOSOR23898-1GEA 39.3 EUR
APOSOR23898-1G
Ethyl-2-amino-4-methylpyrimidine-5-carboxylate 97%
Ethyl-2-amino-4-methylpyrimidine-5-carboxylate
Formel: C₈H₁₁N₃O₂
Molecular Weight: 181.2 g/mol
Schmelzpunkt: 220 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00052622
CAS-Nummer: 81633-29-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR