5-Chloro-1,3-dimethyl-4-(tetrahydro-1H-pyrrol-1-ylsulphonyl)-1H-pyrazole

Lieferant: Apollo Scientific
Achtung

OR23903-1G
APOSOR23903-1GEA 183 EUR
APOSOR23903-1G
5-Chloro-1,3-dimethyl-4-(tetrahydro-1H-pyrrol-1-ylsulphonyl)-1H-pyrazole
5-Chloro-1,3-dimethyl-4-(tetrahydro-1H-pyrrol-1-ylsulphonyl)-1H-pyrazole
Formel: C₉H₁₄ClN₃O₂S
Molecular Weight: 263,744 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00113247
CAS-Nummer: 647825-38-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR