Methyl piperazine-2-carboxylate, N1-CBZ protected 97%
Lieferant: Apollo Scientific
Synonyme:
1-Benzyl 2-methyl piperazine-1,2-dicarboxylate, Piperazine-2-carboxylic acid methyl ester, N1-CBZ protected
OR2391-250MG
APOSOR2391-250MGEA
92.8
EUR
APOSOR2391-250MG
Methyl piperazine-2-carboxylate, N1-CBZ protected 97%
Methyl piperazine-2-carboxylate, N1-CBZ protected
Formel:
C₁₄H₁₈N₂O₄ Lagertemperatur: Raumtemperatur |
MDL:
MFCD04115344 CAS-Nummer: 126937-43-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...