5-Chlor-1,3-dimethyl-1H-pyrazol-4-sulfonamid

Lieferant: Apollo Scientific
Achtung

OR23929-250MG
APOSOR23929-250MGEA 27.7 EUR
APOSOR23929-250MG
5-Chlor-1,3-dimethyl-1H-pyrazol-4-sulfonamid
5-Chlor-1,3-dimethyl-1H-pyrazol-4-sulfonamid
Formel: C₅H₈ClN₃O₂S
Molecular Weight: 209,66 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00105874
CAS-Nummer: 88398-46-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR