4,4'-Dichlorbenzhydrol

Lieferant: Apollo Scientific
Achtung

Synonyme: 4,4'-Dichlorobenzhydryl alcohol, DBH, 4,4'-Dichlorophenyl phenyl carbinol, 4,4'-Dichloro-α-phenylbenzyl alcohol, 4,4'-Dichlorodiphenylmethanol, Bis(4-chlorophenyl)methanol, Bis(4-chlorophenyl)carbinol

OR2394-25G
APOSOR2394-25GEA 66.7 EUR
APOSOR2394-25G
4,4'-Dichlorbenzhydrol
4,4'-Dichlorbenzhydrol
Formel: C₁₃H₁₀Cl₂O
Molecular Weight: 253,13 g/mol
Schmelzpunkt: 91…93 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00000629
CAS-Nummer: 90-97-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR