5-(4-Methylphenyl)cyclohexane-1,3-dione

Lieferant: Apollo Scientific

OR23987-1G OR23987-250MG OR23987-2.5G OR23987-5G
APOSOR23987-1GEA 366 EUR
APOSOR23987-1G APOSOR23987-250MG APOSOR23987-2.5G APOSOR23987-5G
5-(4-Methylphenyl)cyclohexane-1,3-dione
5-(4-Methylphenyl)cyclohexane-1,3-dione
Formel: C₁₃H₁₄O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00111717
CAS-Nummer: 61888-37-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR