5-(4-Methylphenyl)cyclohexane-1,3-dione
Lieferant: Apollo Scientific
OR23987-1G
OR23987-250MG
OR23987-2.5G
OR23987-5G
APOSOR23987-1GEA
366
EUR
APOSOR23987-1G
APOSOR23987-250MG
APOSOR23987-2.5G
APOSOR23987-5G
5-(4-Methylphenyl)cyclohexane-1,3-dione
5-(4-Methylphenyl)cyclohexane-1,3-dione
Formel:
C₁₃H₁₄O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00111717 CAS-Nummer: 61888-37-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...