2,6-Dimethoxybenzaldehyd 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR24023-5G
OR24023-100G
APOSOR24023-5GEA
0
EUR
APOSOR24023-5G
APOSOR24023-100G
2,6-Dimethoxybenzaldehyd 98%
2,6-Dimethoxybenzaldehyde
Formel:
C₉H₁₀O₃ Molecular Weight: 166.18 g/mol Siedepunkt: 285 °C Schmelzpunkt: 96…98 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010862 CAS-Nummer: 3392-97-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...