4-Acetyl-4'-methoxydiphenyl ether
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-(4-(4-Methoxyphenoxy)phenyl)ethanone, 4'-(4-Methoxyphenoxy)acetophenone, 4-Acetyl-4'-methoxydiphenyl ether
OR24033-1G
APOSOR24033-1GEA
46.4
EUR
APOSOR24033-1G
4-Acetyl-4'-methoxydiphenyl ether
4-Acetyl-4'-methoxydiphenyl ether
Formel:
C₁₅H₁₄O₃ Molecular Weight: 242,28 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00052928 CAS-Nummer: 54916-28-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...