4-(5-Oxazolyl)benzonitril

Lieferant: Apollo Scientific
Achtung

OR24050-1G
APOSOR24050-1GEA 65.4 EUR
APOSOR24050-1G
4-(5-Oxazolyl)benzonitril
4-(5-Oxazolyl)benzonitril
Formel: C₁₀H₆N₂O
Molecular Weight: 170,1674 g/mol
Schmelzpunkt: 135…137 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00084987
CAS-Nummer: 87150-13-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR