5-(2-Nitrophenyl)oxazol

Lieferant: Apollo Scientific
Achtung

OR24051-1G OR24051-500MG
APOSOR24051-1GEA 0 EUR
APOSOR24051-1G APOSOR24051-500MG
5-(2-Nitrophenyl)oxazol
5-(2-Nitrophenyl)oxazole
Formel: C₉H₆N₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00085146
CAS-Nummer: 89808-75-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR