N-(Cyanacetyl)morpholin 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Morpholino-3-oxopropanenitrile, 3-(Morpholin-4-yl)-3-oxopropanenitrile
OR24065-1G
APOSOR24065-1GEA
21.9
EUR
APOSOR24065-1G
N-(Cyanacetyl)morpholin 98%
N-(Cyanacetyl)morpholin
Formel:
C₇H₁₀N₂O₂ Schmelzpunkt: 85…88 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00023353 CAS-Nummer: 15029-32-0 EINECS: 239-112-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...