ω-Brom-4-nitroacetophenon 98%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
2-Brom-4'-nitroacetophenon, 4-Nitrophenacylbromid, Brommethyl-p-nitrophenylketon, α-Brom-4-nitroacetophenon
OR24069-25G
APOSOR24069-25GEA
21.9
EUR
APOSOR24069-25G
ω-Brom-4-nitroacetophenon 98%
ω-Brom-4-nitroacetophenon
Formel:
C₈H₆BrNO₃ Schmelzpunkt: 94…101 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007356 CAS-Nummer: 99-81-0 EINECS: 202-789-9 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...