2-Bromo-1-(4-chlorophenyl)-2-(4-methylphenyl)ethanone

Lieferant: Apollo Scientific
Gefahr

OR24072-1G
APOSOR24072-1GEA 174 EUR
APOSOR24072-1G
2-Bromo-1-(4-chlorophenyl)-2-(4-methylphenyl)ethanone
2-Bromo-1-(4-chlorophenyl)-2-(4-methylphenyl)ethanone
Formel: C₁₅H₁₂BrClO
Molecular Weight: 323,62 g/mol
Schmelzpunkt: 76…78 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00052202
CAS-Nummer: 119267-79-7
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR