4-Methylimidazolidine-2-thione
Lieferant: Apollo Scientific
|
Achtung
|
OR24086-1G
OR24086-100MG
OR24086-250MG
OR24086-500MG
OR24086-2.5G
OR24086-5G
APOSOR24086-1GEA
589
EUR
APOSOR24086-1G
APOSOR24086-100MG
APOSOR24086-250MG
APOSOR24086-500MG
APOSOR24086-2.5G
APOSOR24086-5G
4-Methylimidazolidine-2-thione
4-Methylimidazolidine-2-thione
Formel:
C₄H₈N₂S Lagertemperatur: Raumtemperatur |
MDL:
MFCD00121569 CAS-Nummer: 2122-19-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...