4-Methylimidazolidine-2-thione

Lieferant: Apollo Scientific
Achtung

OR24086-1G OR24086-100MG OR24086-250MG OR24086-500MG OR24086-2.5G OR24086-5G
APOSOR24086-1GEA 589 EUR
APOSOR24086-1G APOSOR24086-100MG APOSOR24086-250MG APOSOR24086-500MG APOSOR24086-2.5G APOSOR24086-5G
4-Methylimidazolidine-2-thione
4-Methylimidazolidine-2-thione
Formel: C₄H₈N₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD00121569
CAS-Nummer: 2122-19-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR