2,5-Dichlorbenzonitril 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR24097-5G
APOSOR24097-5GEA
21.9
EUR
APOSOR24097-5G
2,5-Dichlorbenzonitril 98%
2,5-Dichlorbenzonitril
Formel:
C₇H₃Cl₂N Molecular Weight: 172,01 g/mol Schmelzpunkt: 128…132 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00019741 CAS-Nummer: 21663-61-6 EINECS: 244-518-7 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...