4-Chloro-3-nitrobenzenesulphonamide

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Nitro-4-chlorobenzenesulphonamide

OR24156-25G
APOSOR24156-25GEA 21.9 EUR
APOSOR24156-25G
4-Chloro-3-nitrobenzenesulphonamide
4-Chloro-3-nitrobenzenesulphonamide
Formel: C₆H₅ClN₂O₄S
Molecular Weight: 236,633 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00035783
CAS-Nummer: 97-09-6
EINECS: 202-559-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR