2-Chloro-5-nitro-N-(4-pyridyl)benzamide 98+%
Lieferant: Apollo Scientific
Synonyme:
T0070907, 2-Chloro-5-nitro-N-(pyridin-4-yl)benzamide
OR24163-250MG
APOSOR24163-250MGEA
71.2
EUR
APOSOR24163-250MG
2-Chloro-5-nitro-N-(4-pyridyl)benzamide 98+%
2-Chloro-5-nitro-N-(4-pyridyl)benzamide
Formel:
C₁₂H₈ClN₃O₃ Molecular Weight: 277,7 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00121849 CAS-Nummer: 313516-66-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...