tert-Butyl-3-aminobenzoat
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
tert-Butyl 3-aminobenzoate
OR2417-1G
OR2417-25G
OR2417-5G
APOSOR2417-1GEA
125
EUR
APOSOR2417-1G
APOSOR2417-25G
APOSOR2417-5G
tert-Butyl-3-aminobenzoat
tert-Butyl-3-aminobenzoat
Formel:
C₁₁H₁₅NO₂ Schmelzpunkt: 76…78 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00729048 CAS-Nummer: 92146-82-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...