Bis(5-chloro-1,3-dimethyl-1H-pyrazol-4-yl)methanol

Lieferant: Apollo Scientific
Achtung

OR24176-100MG
APOSOR24176-100MGEA 40.6 EUR
APOSOR24176-100MG
Bis(5-chloro-1,3-dimethyl-1H-pyrazol-4-yl)methanol
Bis(5-chloro-1,3-dimethyl-1H-pyrazol-4-yl)methanol
Formel: C₁₁H₁₄Cl₂N₄O
Molecular Weight: 289,161 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00121975
CAS-Nummer: 648409-46-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR