Ethyl-5-amino-1-methyl-1H-pyrazol-4-carboxylat

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl 5-amino-1-methyl-1H-pyrazole-4-carboxylate

OR24192-1G
APOSOR24192-1GEA 21.9 EUR
APOSOR24192-1G
Ethyl-5-amino-1-methyl-1H-pyrazol-4-carboxylat
Ethyl-5-amino-1-methyl-1H-pyrazol-4-carboxylat
Formel: C₇H₁₁N₃O₂
Molecular Weight: 169.18 g/mol
Schmelzpunkt: 96…100 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00051652
CAS-Nummer: 31037-02-2
EINECS: 31037-02-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR