(±)-1,2,3,4-Tetrahydro-1-naphthalinol 95+%
Lieferant: Apollo Scientific
Synonyme:
ac-alpha-Tetralol, 1,2,3,4-Tetrahydronaphthalen-1-ol, 1,2,3,4-Tetrahydro-1-naphthol
OR24205-25G
APOSOR24205-25GEA
60.9
EUR
APOSOR24205-25G
(±)-1,2,3,4-Tetrahydro-1-naphthalinol 95+%
(±)-1,2,3,4-Tetrahydro-1-naphthol
Formel:
C₁₀H₁₂O Molecular Weight: 148,2 g/mol Siedepunkt: 102…104 °C Dichte: 1,09 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001739 CAS-Nummer: 529-33-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...