3-Nitro-1H-pyrazol
Lieferant: Apollo Scientific
|
Achtung
|
OR24208-25G
APOSOR24208-25GEA
21.9
EUR
APOSOR24208-25G
3-Nitro-1H-pyrazol
3-Nitro-1H-pyrazole
Formel:
C₃H₃N₃O₂ Molecular Weight: 113,08 g/mol Schmelzpunkt: 173…175 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00238787 CAS-Nummer: 26621-44-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...