6-Chloro[1,2,4]triazolo[4,3-b]pyridazine
Lieferant: Apollo Scientific
|
Achtung
|
OR24235-250MG
APOSOR24235-250MGEA
21.9
EUR
APOSOR24235-250MG
6-Chloro[1,2,4]triazolo[4,3-b]pyridazine
6-Chloro[1,2,4]triazolo[4,3-b]pyridazine
Formel:
C₅H₃ClN₄ Schmelzpunkt: 202…210 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00219047 CAS-Nummer: 28593-24-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...