Methyl-3-amino-5-phenyl-2-thiophencarboxylat
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl 3-amino-5-phenylthiophene-2-carboxylate
OR24261-25G
APOSOR24261-25GEA
145
EUR
APOSOR24261-25G
Methyl-3-amino-5-phenyl-2-thiophencarboxylat
Methyl-3-amino-5-phenyl-2-thiophencarboxylat
Formel:
C₁₂H₁₁NO₂S Molecular Weight: 233.2862 g/mol Schmelzpunkt: 145…149 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00068161 CAS-Nummer: 100063-22-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...