Ethyl 1-methyl-5-(1H-pyrrol-1-yl)-1H-pyrazole-4-carboxylate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-[4-(Ethoxycarbonyl)-1-methyl-1H-pyrazol-5-yl]-1H-pyrrole, 4-(Ethoxycarbonyl)-1-methyl-5-(1H-pyrrol-1-yl)-1H-pyrazole
OR24264-250MG
APOSOR24264-250MGEA
128
EUR
APOSOR24264-250MG
Ethyl 1-methyl-5-(1H-pyrrol-1-yl)-1H-pyrazole-4-carboxylate
Ethyl 1-methyl-5-(1H-pyrrol-1-yl)-1H-pyrazole-4-carboxylate
Formel:
C₁₁H₁₃N₃O₂ Molecular Weight: 219,24 g/mol Schmelzpunkt: 87 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00052539 CAS-Nummer: 175137-01-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...