Ethyl 5-propyl-1H-pyrazole-4-carboxylate

Lieferant: Apollo Scientific
Achtung

OR24284-500MG
APOSOR24284-500MGEA 21.9 EUR
APOSOR24284-500MG
Ethyl 5-propyl-1H-pyrazole-4-carboxylate
Ethyl 5-propyl-1H-pyrazole-4-carboxylate
Formel: C₉H₁₄N₂O₂
Molecular Weight: 182,22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01570118
CAS-Nummer: 123374-28-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR