2-Acetyl-3-amino-5-tert-butylthiophen

Lieferant: Apollo Scientific
Achtung

OR24302-100MG
APOSOR24302-100MGEA 29 EUR
APOSOR24302-100MG
2-Acetyl-3-amino-5-tert-butylthiophen
2-Acetyl-3-amino-5-tert-butylthiophen
Formel: C₁₀H₁₅NOS
Lagertemperatur: Kühlschrank
MDL: MFCD00068131
CAS-Nummer: 175137-06-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR