2H-Thieno[3,2-d][1,3]oxazine-2,4(1H)-dione

Lieferant: Apollo Scientific
Achtung

Synonyme: 1,4-Dihydro-2,4-dioxo-2H-thieno[3,2-d][1,3]oxazine, 1H-Thieno[3,2-d][1,3]oxazine-2,4-dione

OR24307-250MG
APOSOR24307-250MGEA 34.8 EUR
APOSOR24307-250MG
2H-Thieno[3,2-d][1,3]oxazine-2,4(1H)-dione
2H-Thieno[3,2-d][1,3]oxazine-2,4(1H)-dione
Formel: C₆H₃NO₃S
Molecular Weight: 169,158 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00106648
CAS-Nummer: 78756-28-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR